Assay Design



Gene Transcripts

Gene Symbol
GLTPD2
Entrez Gene ID
607478
Gene Name
glycolipid transfer protein domain containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 31781105 - 31784373 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005619933.3 XP_005619990.1 4 - 4 660 84
XM_844159.4 XP_849252.1 3 - 3 760 84

View Full Product Details