Assay Design



Gene Transcripts

Gene Symbol
CKM
Entrez Gene ID
476435
Gene Name
creatine kinase, M-type
Gene Aliases
CPK-M, M-CK
Chromosome Location
Chr.1: 110210158 - 110218937 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313776.1 NP_001300705.1 3 - 4 419 75

View Full Product Details