Assay Design



Gene Transcripts

Gene Symbol
INHBB
Entrez Gene ID
483880
Gene Name
inhibin subunit beta B
Gene Aliases
-
Chromosome Location
Chr.19: 29765697 - 29768485 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631933.1 XP_005631990.1 1 - 1 117 83

View Full Product Details