Assay Design



Gene Transcripts

Gene Symbol
MYC
Entrez Gene ID
403924
Gene Name
MYC proto-oncogene, bHLH transcription factor
Gene Aliases
C-MYC
Chromosome Location
Chr.13: 25200772 - 25205309 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003246.2 NP_001003246.2 - 483 61
GenBank mRNA DN374130.1 - - 483 61
X95367.1 - - 60 61

View Full Product Details