Assay Design



Gene Transcripts

Gene Symbol
NOVA2
Entrez Gene ID
612213
Gene Name
NOVA alternative splicing regulator 2
Gene Aliases
-
Chromosome Location
Chr.1: 109657693 - 109683277 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116591.2 XP_013972066.1 3 - 3 538 177

View Full Product Details