Assay Design
Gene Transcripts
- Gene Symbol
- PAX1
- Entrez Gene ID
-
485743
- Gene Name
- paired box 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.24: 1742570 - 1752593 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_542866.6 |
XP_542866.3 |
2 - 3 |
903 |
97 |
View Full Product Details