Assay Design
Gene Transcripts
- Gene Symbol
- EIF4EBP1
- Entrez Gene ID
-
475590
- Gene Name
- eukaryotic translation initiation factor 4E binding protein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 27389850 - 27410154 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_549448.5 |
XP_549448.2 |
- |
104 |
157 |
View Full Product Details