Assay Design



Gene Transcripts

Gene Symbol
FEN1
Entrez Gene ID
476063
Gene Name
flap structure-specific endonuclease 1
Gene Aliases
-
Chromosome Location
Chr.18: 54680627 - 54686502 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631652.3 XP_005631709.1 2 - 2 1041 164
XM_022405811.1 XP_022261519.1 2 - 2 1071 164
XM_533271.6 XP_533271.1 2 - 2 1129 164

View Full Product Details