Assay Design



Gene Transcripts

Gene Symbol
ESRRB
Entrez Gene ID
490798
Gene Name
estrogen related receptor beta
Gene Aliases
-
Chromosome Location
Chr.8: 49253877 - 49424148 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116090.2 XP_013971565.1 6 - 6 1128 140
XM_022422243.1 XP_022277951.1 6 - 6 1361 140
XM_022422244.1 XP_022277952.1 6 - 6 1441 140
XM_547920.6 XP_547920.4 6 - 6 1280 140

View Full Product Details