Assay Design
Gene Transcripts
- Gene Symbol
- POU4F2
- Entrez Gene ID
-
482637
- Gene Name
- POU class 4 homeobox 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 45030818 - 45034446 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_848360.2 |
XP_853453.1 |
2 - 2 |
671 |
102 |
View Full Product Details