Assay Design



Gene Transcripts

Gene Symbol
LHX1
Entrez Gene ID
100856649
Gene Name
LIM homeobox 1
Gene Aliases
-
Chromosome Location
Chr.9: 36760890 - 36768345 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116734.2 XP_013972209.1 2 - 2 1170 122

View Full Product Details