Assay Design



Gene Transcripts

Gene Symbol
SACS
Entrez Gene ID
477339
Gene Name
sacsin molecular chaperone
Gene Aliases
-
Chromosome Location
Chr.25: 15207959 - 15297483 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005635545.3 XP_005635602.2 - 14013 132
XM_014107391.2 XP_013962866.1 - 14040 132
XM_014107392.2 XP_013962867.1 - 13259 132

View Full Product Details