Assay Design
Gene Transcripts
- Gene Symbol
- CHSY1
- Entrez Gene ID
-
488704
- Gene Name
- chondroitin sulfate synthase 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 39844227 - 39916484 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_545821.6 |
XP_545821.3 |
2 - 3 |
827 |
132 |
View Full Product Details