Assay Design



Gene Transcripts

Gene Symbol
PDP2
Entrez Gene ID
489771
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 2
Gene Aliases
-
Chromosome Location
Chr.5: 82421531 - 82428600 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620865.3 XP_005620922.1 2 - 2 1010 95
XM_005620866.2 XP_005620923.1 2 - 2 913 95
XM_022419395.1 XP_022275103.1 2 - 2 905 95
XM_546891.6 XP_546891.2 2 - 2 1005 95

View Full Product Details