Assay Design
Gene Transcripts
- Gene Symbol
- GATAD1
- Entrez Gene ID
-
482301
- Gene Name
- GATA zinc finger domain containing 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 18043857 - 18054340 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_539418.5 |
XP_539418.3 |
2 - 3 |
493 |
162 |
View Full Product Details