Assay Design
Gene Transcripts
- Gene Symbol
- MRPL32
- Entrez Gene ID
-
475869
- Gene Name
- mitochondrial ribosomal protein L32
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 7212604 - 7218051 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_533077.6 |
XP_533077.1 |
- |
510 |
171 |
View Full Product Details