Assay Design



Gene Transcripts

Gene Symbol
DNAJC30
Entrez Gene ID
489805
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
Gene Aliases
-
Chromosome Location
Chr.6: 6595233 - 6596440 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620967.3 XP_005621024.1 - 331 147

View Full Product Details