Assay Design
Gene Transcripts
- Gene Symbol
- TADA1
- Entrez Gene ID
-
480090
- Gene Name
- transcriptional adaptor 1
- Gene Aliases
-
TADA1L
- Chromosome Location
-
Chr.7: 31513289 - 31529891 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_537213.6 |
XP_537213.2 |
5 - 6 |
770 |
132 |
View Full Product Details