Assay Design
Gene Transcripts
- Gene Symbol
- NLRP6
- Entrez Gene ID
-
483394
- Gene Name
- NLR family pyrin domain containing 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 25433481 - 25440703 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_540513.5 |
XP_540513.3 |
4 - 4 |
1388 |
76 |
View Full Product Details