Assay Design
Gene Transcripts
- Gene Symbol
- GPR26
- Entrez Gene ID
-
486926
- Gene Name
- G protein-coupled receptor 26
- Gene Aliases
-
-
- Chromosome Location
-
Chr.28: 33152088 - 33182638 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_544056.5 |
XP_544056.2 |
1 - 1 |
697 |
134 |
View Full Product Details