Assay Design
Gene Transcripts
- Gene Symbol
- MBLAC2
- Entrez Gene ID
-
488914
- Gene Name
- metallo-beta-lactamase domain containing 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 18467948 - 18485700 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_546031.6 |
XP_546031.1 |
- |
970 |
130 |
View Full Product Details