Assay Design



Gene Transcripts

Gene Symbol
TSEN54
Entrez Gene ID
483315
Gene Name
tRNA splicing endonuclease subunit 54
Gene Aliases
-
Chromosome Location
Chr.9: 5009468 - 5016513 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022422730.1 XP_022278438.1 - 498 131
XM_540434.6 XP_540434.3 - 498 131

View Full Product Details