Assay Design
Gene Transcripts
- Gene Symbol
- HSPE1
- Entrez Gene ID
-
478855
- Gene Name
- heat shock 10kDa protein 1 (chaperonin 10)
- Gene Aliases
-
MOB4, MOBKL3, PREI3
- Chromosome Location
-
Chr.37: 7020507 - 7063678 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_536018.6 |
XP_536018.2 |
1 - 2 |
160 |
151 |
View Full Product Details