Assay Design
Gene Transcripts
- Gene Symbol
- THAP9
- Entrez Gene ID
-
487834
- Gene Name
- THAP domain containing 9
- Gene Aliases
-
-
- Chromosome Location
-
Chr.32: 6852057 - 6875660 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_544956.6 |
XP_544956.2 |
1 - 2 |
142 |
61 |
View Full Product Details