Assay Design



Gene Transcripts

Gene Symbol
SUCNR1
Entrez Gene ID
485719
Gene Name
succinate receptor 1
Gene Aliases
-
Chromosome Location
Chr.23: 46415153 - 46424303 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022408748.1 XP_022264456.1 2 - 2 2117 158

View Full Product Details