Assay Design



Gene Transcripts

Gene Symbol
THAP6
Entrez Gene ID
487811
Gene Name
THAP domain containing 6
Gene Aliases
-
Chromosome Location
Chr.32: 94055 - 111365 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005639055.2 XP_005639112.1 4 - 5 582 153
XM_005639056.3 XP_005639113.1 3 - 4 740 153
XM_005639057.3 XP_005639114.1 3 - 4 449 153
XM_843217.5 XP_848310.1 4 - 5 841 153

View Full Product Details