Assay Design



Gene Transcripts

Gene Symbol
ADRB3
Entrez Gene ID
442979
Gene Name
adrenoceptor beta 3
Gene Aliases
B3AR, GPCR
Chromosome Location
Chr.16: 27445601 - 27446796 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003377.1 NP_001003377.1 - 274 94
GenBank mRNA AF200597.1 - - 274 94

View Full Product Details