Assay Design



Gene Transcripts

Gene Symbol
B3GAT1
Entrez Gene ID
489265
Gene Name
beta-1,3-glucuronyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.5: 741231 - 747641 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031634.1 NP_001026804.1 - 627 77
GenBank mRNA AJ888975.1 - - 627 77

View Full Product Details