Assay Design



Gene Transcripts

Gene Symbol
KRT12
Entrez Gene ID
100034663
Gene Name
keratin 12
Gene Aliases
-
Chromosome Location
Chr.9: 21823976 - 21829557 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082421.1 NP_001075890.1 - 1417 104
GenBank mRNA EF382361.1 - - 1417 104

View Full Product Details