Assay Design



Gene Transcripts

Gene Symbol
GHSR
Entrez Gene ID
100101479
Gene Name
growth hormone secretagogue receptor
Gene Aliases
-
Chromosome Location
Chr.34: 36691132 - 36694304 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099945.1 NP_001093415.1 1 - 2 752 129
GenBank mRNA EF536345.1 - 1 - 2 752 129

View Full Product Details