Assay Design



Gene Transcripts

Gene Symbol
PHB
Entrez Gene ID
480547
Gene Name
prohibitin
Gene Aliases
-
Chromosome Location
Chr.9: 25525711 - 25536722 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110179.2 NP_001103649.1 - 153 161
GenBank mRNA DN877880.1 - 1 - 1 153 161
EU188843.1 - 1 - 1 109 161

View Full Product Details