Assay Design



Gene Transcripts

Gene Symbol
RSPO1
Entrez Gene ID
608179
Gene Name
R-spondin 1
Gene Aliases
-
Chromosome Location
Chr.15: 4857887 - 4876077 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130838.1 NP_001124310.1 4 - 5 626 101
XM_005628844.3 XP_005628901.1 5 - 6 1026 101
XM_022427669.1 XP_022283377.1 5 - 6 968 101
GenBank mRNA EU038071.1 - 4 - 5 626 101

View Full Product Details