Assay Design



Gene Transcripts

Gene Symbol
TAS2R10
Entrez Gene ID
100271734
Gene Name
taste 2 receptor member 10
Gene Aliases
CAFA-T2R10
Chromosome Location
Chr.27: 34876090 - 34877058 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145497.1 NP_001138969.1 1 - 1 842 99

View Full Product Details