Assay Design



Gene Transcripts

Gene Symbol
KRT19
Entrez Gene ID
100685481
Gene Name
keratin 19
Gene Aliases
-
Chromosome Location
Chr.9: 21240778 - 21245039 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253742.1 NP_001240671.1 - 997 67
GenBank mRNA DN879929.1 - - 558 67
EU107521.1 - - 52 67

View Full Product Details