Assay Design



Gene Transcripts

Gene Symbol
KLK10
Entrez Gene ID
484351
Gene Name
kallikrein related peptidase 10
Gene Aliases
-
Chromosome Location
Chr.1: 105860406 - 105864238 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197158.1 NP_001184087.1 4 - 5 683 78
XM_005615761.3 XP_005615818.1 5 - 6 818 78
GenBank mRNA GQ303578.1 - 4 - 5 683 78

View Full Product Details