Assay Design



Gene Transcripts

Gene Symbol
TMED7
Entrez Gene ID
481436
Gene Name
transmembrane p24 trafficking protein 7
Gene Aliases
-
Chromosome Location
Chr.11: 5342265 - 5356828 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204338.1 NP_001191267.1 - 384 79

View Full Product Details