Assay Design



Gene Transcripts

Gene Symbol
RFX8
Entrez Gene ID
100683612
Gene Name
regulatory factor X8
Gene Aliases
-
Chromosome Location
Chr.10: 41464882 - 41521157 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270887.1 NP_001257816.1 6 - 7 610 114
XM_022423975.1 XP_022279683.1 9 - 10 847 114

View Full Product Details