Assay Design



Gene Transcripts

Gene Symbol
RFX8
Entrez Gene ID
100683612
Gene Name
regulatory factor X8
Gene Aliases
-
Chromosome Location
Chr.10: 41464882 - 41521157 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270887.1 NP_001257816.1 8 - 9 867 104
XM_022423975.1 XP_022279683.1 11 - 12 1104 104

View Full Product Details