Assay Design



Gene Transcripts

Gene Symbol
IL9
Entrez Gene ID
100682918
Gene Name
interleukin 9
Gene Aliases
-
Chromosome Location
Chr.11: 23808919 - 23814085 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003431593.3 XP_003431641.1 3 - 4 190 70

View Full Product Details