Assay Design



Gene Transcripts

Gene Symbol
Sec22b
Entrez Gene ID
100689313
Gene Name
SEC22 vesicle trafficking protein homolog B (S. cerevisiae)
Gene Aliases
ERS-24, Ers24, I79_012586
Chromosome Location
Chr.Un|NW_003614146: 575565 - 595729 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246746.1 NP_001233675.1 3 - 4 431 81
GenBank mRNA JI875226.1 - 3 - 4 486 81
JP060503.1 - 3 - 4 486 81
U91742.1 - 3 - 4 431 81

View Full Product Details