Assay Design



Gene Transcripts

Gene Symbol
Lhb
Entrez Gene ID
100689384
Gene Name
luteinizing hormone beta
Gene Aliases
I79_016647
Chromosome Location
Chr.Un|NW_003614570: 327639 - 328567 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246810.2 NP_001233739.1 2 - 3 189 98
GenBank mRNA AB248598.1 - 2 - 3 191 98

View Full Product Details