Assay Design



Gene Transcripts

Gene Symbol
Gna11
Entrez Gene ID
100689065
Gene Name
guanine nucleotide binding protein (G protein), alpha 11 (Gq class)
Gene Aliases
I79_013821
Chromosome Location
Chr.Un|NW_003614244: 352484 - 357164 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244051.1 NP_001230980.1 5 - 6 890 97
GenBank mRNA DQ202706.1 - 5 - 6 890 97
JI862434.1 - 5 - 6 752 97
JP039111.1 - 5 - 6 753 97

View Full Product Details