Assay Design



Gene Transcripts

Gene Symbol
St8sia4
Entrez Gene ID
100689217
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4
Gene Aliases
I79_014283, PST-1, Pst1, SIAT8-D, SIAT8D, ST8SiaIV
Chromosome Location
Chr.Un|NW_003614294: 143025 - 224209 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244250.1 NP_001231179.1 4 - 5 1090 74
GenBank mRNA Z46801.1 - 4 - 5 1090 74

View Full Product Details