Assay Design



Gene Transcripts

Gene Symbol
Mt2
Entrez Gene ID
100689479
Gene Name
metallothionein 2
Gene Aliases
MT-2, MT-II
Chromosome Location
Chr.Un|NW_003613984: 785539 - 786096 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244577.1 NP_001231506.1 1 - 2 25 87

View Full Product Details