Assay Design



Gene Transcripts

Gene Symbol
Gpx3
Entrez Gene ID
100764638
Gene Name
glutathione peroxidase 3 (plasma)
Gene Aliases
I79_005447
Chromosome Location
Chr.Un|NW_003613733: 925580 - 933676 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256815.1 NP_001243744.1 1 - 2 203 75

View Full Product Details