Assay Design



Gene Transcripts

Gene Symbol
Zscan21
Entrez Gene ID
100773934
Gene Name
zinc finger and SCAN domain containing 21
Gene Aliases
I79_018693
Chromosome Location
Chr.Un|NW_003614875: 139237 - 149348 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510415.2 XP_003510463.1 3 - 4 811 74
XM_007651030.1 XP_007649220.1 3 - 4 749 74
XM_007651031.1 XP_007649221.1 3 - 4 951 74
XM_007651032.1 XP_007649222.1 3 - 4 792 74

View Full Product Details