Assay Design



Gene Transcripts

Gene Symbol
Ptrh1
Entrez Gene ID
100760505
Gene Name
peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae)
Gene Aliases
I79_014783
Chromosome Location
Chr.Un|NW_003614338: 353432 - 354759 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647735.1 XP_007645925.1 2 - 3 351 80

View Full Product Details