Assay Design



Gene Transcripts

Gene Symbol
Il17a
Entrez Gene ID
100750422
Gene Name
interleukin 17A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614654: 428386 - 431477 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509128.1 XP_003509176.1 2 - 3 238 81

View Full Product Details