Assay Design



Gene Transcripts

Gene Symbol
En1
Entrez Gene ID
103163841
Gene Name
engrailed homeobox 1
Gene Aliases
I79_005518
Chromosome Location
Chr.Un|NW_003613737: 1194640 - 1204878 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640208.1 XP_007638398.1 2 - 3 372 64

View Full Product Details