Assay Design



Gene Transcripts

Gene Symbol
Epm2aip1
Entrez Gene ID
100770168
Gene Name
EPM2A (laforin) interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614270: 490980 - 494841 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506672.2 XP_003506720.1 1 - 1 1215 85

View Full Product Details